missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Catalog Number p-4510239
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3087-36-3
C8H20O4Ti
228.15
JMXKSZRRTHPKDL-UHFFFAOYSA-N
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
76524
ethanolate;titanium(4+)
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
3087-36-3
C8H20O4Ti
228.15
JMXKSZRRTHPKDL-UHFFFAOYSA-N
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
76524
ethanolate;titanium(4+)
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 150.0°C to 152.0°C (10.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula Ti(OC2H5)4
Refractive Index 1.5050 to 1.5070
Quantity 50 g
Beilstein 01,III,1338
Specific Gravity 1.09
Solubility Information Solubility in water: hydrolysis.
Viscosity 60-80 mPa.s (20°C)
Formula Weight 228.15
Percent Purity 33-35% TiO2
Physical Form Liquid
Chemical Name or Material Titanium(IV) ethoxide
Show More Show Less
compliance-icons
Product Identifier
  • Titanium(IV) ethoxide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'