missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | >3000°C |
Melting Point | 1700°C |
Physical Form | Lump |
Quantity | 500 g |
Assay Percent Range | 99.9%, metal basis |
Solubility Information | Soluble in concentrated sulfuric acid,concentrated hydrofluoric acid and alkali. Insoluble in water,diluted sulfuric acid,hydrochloric acid,nitric acid and organic solvents. |
Formula Weight | 63.9 |
Density | 4.95 |
Chemical Name or Material | Titanium(II) Oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'