missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4408021
Click to view available options
Quantity:
5 g
100 g
500 g
2 x 1 kg
This item is not returnable. View return policy
7704-98-5
H2Ti
MFCD00151301
This item is not returnable. View return policy
7704-98-5
H2Ti
MFCD00151301

Widley used as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics.

Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 450°C (decomposition)
Physical Form Powder
Quantity 5 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in water.
Formula Weight 49.92
Sensitivity Air and moisture sensitive
Density 3.9
Chemical Name or Material Titanium(II) hydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'