missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number 11305878
Click to view available options
Quantity:
100 x 100 mm
200 x 200 mm
200 x 1000 mm
This item is not returnable. View return policy
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
This item is not returnable. View return policy
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]

Form: Approx. 40g/100 x 100mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Dark Gray foil
Density 4.506g/cm3
Assay Percent Range 99.7% (Metals basis)
Quantity 100 x 100 mm
Odor Odorless
Form Foil
Chemical Name or Material Titanium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'