missing translation for 'onlineSavingsMsg'
Learn More

Titanium, -100 Mesh, 99.5%, Powder, ACROS Organics™

Brand:  Acros Organics 421011000

Code : 29

Additional Details : CAS Number : 7440-32-6 Weight : 0.10000kg Transport : UN number : 2546 Chem class : 4.2 Pack group : II

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10035990

  • £57.90 / 100g


Estimated Shipment
13-06-2018
Add to basket

Specifications

Chemical Identifiers

Titanium
7440-32-6
99.5%
Ti
MFCD00011264
-100 Mesh
Fine Powder
Ti
47.88

Specifications

Titanium
4.51
-100 Mesh
4.5100g/mL
99.5%
Ti
47.88
07,368; 08,481; 09,466; 11,526; 15,315
Plastic bottle
Gray
3287.0°C
237°C
100g
7440-32-6
Fine Powder
99.5%
Ti
MFCD00011264
15, 9626
0.5% max.
1660.0°C
Videos
SDS