Learn More
TIS11D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561513
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MSTTLLSAFY DVDFLCKTEK SLANLNLNNM LDKKAVGTPV AAAPSSGFAP GFLRRHSASN LHALAHPAPS PGSCSPKF Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
TIS11D Polyclonal antibody specifically detects TIS11D in Human samples. It is validated for Immunohistochemistry.Specifications
TIS11D | |
Polyclonal | |
Unconjugated | |
ZFP36L2 | |
Brf2; Butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36L2; mRNA decay activator protein ZFP36L2; Protein TIS11D; RNF162C; TIS11D; TISD; TPA-induced sequence 11d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; ZFP36L2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P47974 | |
ZFP36L2 | |
Recombinant protein corresponding to Human TIS11D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only