Learn More
Thermo Scientific™ Automation Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms with 384-Channel Disposable Tip Head
Increase pipetting performance with Thermo Scientific™ Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand:  Thermo Scientific™ 947-261
UNSPSC : 41121608
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12914127
  • £2367.04 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Description
Description
  • Available in nonsterile and sterile
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
30μL
Sterile
384 Tips/Rack, 10 Racks/Case, 5 Packs/Case
Agilent Bravo
No
Clear
Bravo and VPrep Automated Liquid Handling Platforms
Documentation
Technical Documents