Thermo Scientific™ Automation Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms with 384-Channel Disposable Tip Head

Increase pipetting performance with Thermo Scientific™ Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 946-261

UNSPSC: 41121608

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Code. 12904127

Quantity Price
1 £ 2367.04 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Color Clear
Volume (Metric) 10μL
Sterility Sterile
For Use With (Equipment) Bravo and VPrep Automated Liquid Handling Platforms
Packaging 384 Tips/Rack, 10 Racks/Case, 5 Packs/Case
Filtered No
Product Line Agilent Bravo

  • Available in nonsterile and sterile
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)

Documentation

Technical documents