missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiotidine, Tocris Bioscience™
Potent, selective H2 antagonist
Brand: Tocris (Bio-Techne) 082610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Potent histamine H2-receptor antagonist with negligible activity against H1- and H3- receptors.Chemical Identifiers
69014-14-8 | |
312.414 | |
tiotidine, 3h tiotidine, tiotidine usan:inn, tiotidinum inn-latin, unii-ezu9aiz69m, tiotidina inn-spanish, 3h-tiotidine, ezu9aiz69m, 2-cyano-1-2-2-diaminomethylene amino-4-thiazolyl methyl thio ethyl-3-methylguanidine, dsstox_cid_26341 | |
1-cyano-3-[2-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-2-methylguanidine |
C10H16N8S2 | |
YDDXVAXDYKBWDX-UHFFFAOYSA-N | |
50287 | |
CN=C(NCCSCC1=CSC(=N1)N=C(N)N)NC#N |
Specifications
Tiotidine | |
69014-14-8 | |
tiotidine, 3h tiotidine, tiotidine usan:inn, tiotidinum inn-latin, unii-ezu9aiz69m, tiotidina inn-spanish, 3h-tiotidine, ezu9aiz69m, 2-cyano-1-2-2-diaminomethylene amino-4-thiazolyl methyl thio ethyl-3-methylguanidine, dsstox_cid_26341 | |
CN=C(NCCSCC1=CSC(=N1)N=C(N)N)NC#N | |
312.414 | |
312.41 |
10mg | |
C10H16N8S2 | |
YDDXVAXDYKBWDX-UHFFFAOYSA-N | |
1-cyano-3-[2-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-2-methylguanidine | |
50287 | |
>99% |