Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Comparator Iron Vario TPTZ Test, 0.1 to 1.8mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand: Lovibond™ 147470
Code : NEW
Additional Details : Weight : 0.72700kg
Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
- The Iron disc supplied measures from 0.1 to 1.8mg/L with the following steps: 0.1 / 0.2 / 0.3 / 0.4 / 0.5 / 0.6 / 0.7 / 0.8 / 0.9 / 1.0 / 1.1 / 1.2 / 1.3 / 1.4 / 1.5 / 1.6 / 1.7 / 1.8
Specifications
Water Quality Testing | |
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests | |
Iron TPTZ Test |
Checkit™ Comparator | |
0.1 to 1.8mg/L |