missing translation for 'onlineSavingsMsg'
Learn More

Tin, Technical, Metal, Granulated, Fisher Chemical™

Click to view available options
Quantity:
100g
500g
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
Boiling Point 2270°C
Color Silver
Melting Point 231.9°C
Quantity 100g
Formula Weight 118.69g/mol
Physical Form Solid
Packaging Plastic powder jar
Vapor Pressure 1mmHg 1492
Form Solid
Product Identifier
  • Tin