Alfa Aesar™ Tin, Oil based standard solution, Specpure™, Sn 5000μg/g


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 042450.18

UNSPSC: 41116107

Code: NEW

Additional Details:
Additional Details: Weight: 0.05000kg


Quantity 50g

Product Code. 15456475

Quantity Price
1 £ 66.78 / 50g
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Tin, Oil based standard solution
Name Note Sn 5000μg/g
Purity Grade Specpure™
MDL Number MFCD00133862

Specification

Name Note Sn 5000μg/g
MDL Number MFCD00133862
Chemical Name or Material Tin, Oil based standard solution
Quantity 50g
Physical Form Liquid
Purity Grade Specpure™
Documentation

Technical documents