missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051467
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
132951-93-0
MFCD00145408
This item is not returnable. View return policy
132951-93-0
MFCD00145408

Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼130°C (decomposition)
Boiling Point 131°C (1.6 mmHg)
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters.
Formula Weight 414.15
Percent Purity 98%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide isopropanol adduct

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'