missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) fluoride, 99% (metals basis), Thermo Scientific Chemicals
56.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
Description
It is used as Friedel-Crafts catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 705°C (sublimation) |
Physical Form | Crystalline |
Quantity | 2 g |
UN Number | UN2923 |
Merck Index | 14,8776 |
Formula Weight | 194.68 |
Percent Purity | 99% |
Sensitivity | Moisture sensitive |
Assay | (metals basis) |
Chemical Name or Material | Tin(IV) fluoride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'