Learn More
Tin(IV) chloride, anhydrous, 98% (metals basis), Thermo Scientific Chemicals
Description
Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which find applications as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in the presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 114°C |
Melting Point | -33°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 g |
Linear Formula | SnCl4 |
UN Number | UN1827 |
Merck Index | 14,8774 |
Solubility Information | Soluble in alcohol,CCl4,benzene,toluene,chloroform,carbon terachloride,gasoline,carbon disulfide and acetone; Soluble in water with much heat evolution. |
Formula Weight | 260.5 |
Show More |
RUO – Research Use Only