missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) sulfate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 360.0°C |
Color | White |
Physical Form | Powder, Crystals and/or Lumps |
Quantity | 100 g |
Assay Percent Range | 94% min. (Iodimetry) |
Linear Formula | SnSO4 |
Merck Index | 15,8917 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in diluted sulfuric acid |
IUPAC Name | tin(2+);sulfate |
Formula Weight | 214.75 |
Show More |
Safety and Handling
Product Identifier
- Tin(II) sulfate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'