missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals
31.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 43°C to 46°C |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 98.0-103.0% |
Solubility Information | Soluble in dilute and concentrated hydrochloric acid,alcohol,ethyl acetate,glacial acetic acid,and sodium hydroxide solution. Decomposes in excess water. |
Formula Weight | 225.65 (189.62 Anhydrous) |
Grade | ACS Reagent |
Sensitivity | Air Sensitive |
Density | 2.71 g/mL |
Chemical Name or Material | Tin(II) chloride dihydrate, For analysis ACS |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'