Learn More
TIMP-1 Ab-2 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TIMP-1 Ab-2, Mouse Monoclonal Antibody.
£121.38 - £674.00
Specifications
Antigen | TIMP-1 Ab-2 |
---|---|
Clone | 102D1 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12661737
|
Epredia
MS-608-B1 |
Concentrated | Biotin | 500 μL |
£470.00
500µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11868731
|
Thermo Scientific
MS608PABX |
Concentrated | Unconjugated | 200 μL |
£674.00
200µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12681737
|
Epredia
MS-608-P |
Concentrated | Unconjugated | 1 mL |
£600.00
1mL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12601747
|
Epredia
MS-608-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12671737
|
Epredia
MS-608-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621747
|
Thermo Scientific
MS-608-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12611747
|
Epredia
MS-608-P1 |
Concentrated | Unconjugated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691737
|
Epredia
MS-608-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651747
|
Epredia
MS-608-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Tissue inhibitor of metalloproteinases-1 (TIMP-1) and Tissue inhibitor of metalloproteinases-2 (TIMP-2) are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-1 forms a complex of 1:1 stoichiometry with activated interstitial collagenase, activated stromelysin, active form of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A), and latent and active forms 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Host Species: Mouse
Clone: 102D1
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human TIMP-1
Molecular Weight: 28.5kDa
Positive Control: Conditioned, serum-free medium from (TPA-treated) human fetal lung cell line HFL-1; Bladder, breast and ovarian CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
TIMP-1 Ab-2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Proteases And Inhibitors | |
Recombinant human TIMP-1 | |
Primary |
102D1 | |
Monoclonal | |
RUO | |
Human | |
IgG1 |