missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Time Switch with LCD display

Programs easily with two buttons. Azpack™ Time Switch with LCD Display has a minimum switch time of only one minute.

Brand:  Azpack™ 12349379

Code : 03

Additional Details : Weight : 0.11340kg

Product Code. 12349379

  • £115.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • 7 day slimline digital plug-in time controller for 13A Resistive(3kW)
  • Inductive 6A 230V AC
  • Individual day,every day, every weekday or weekend programme selections over a 7 day period.
  • Six programmes are available with 42 ON & OFF switchings per week
  • Top side display for ease of viewing when in use.
  • Simple two button programming.
  • Shortest switching time 1 minute.
  • Manual overide button.
  • Optional random switching facility.
  • 1000 hours rechargeable memory back up.
  • Controls 500W of fluorescent or low energy lighting and up to a 1hp motor.
  • Can also be used with up to 1500W of filament lighting.
  • Cannot be used with discharge lighting(e.g. SON and metal Halide)
  • Operating temperature -10 to 40°C
  • Battery factory fitted.
  • Conforms to 73/23/EEC, 89/336/EEC
  • Dimesions: (L x W x H) 65 x 60 x 81mm
Specifications

Specifications

Time Switch
LCD
65 x 60 x 81 mm
230V, 50Hz Single phase supplies
73/23/EEC, 89/336/EEC
7 days
1 min.
Manual override, optional random switching facility
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Do not use with discharge lighting (e.g. SON and metal Halide)