Learn More
TIM-4 Monoclonal Antibody (54 (RMT4-54)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5866-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 54 (RMT4-54) recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC's and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells. Applications Reported: This 54 (RMT4-54) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 54 (RMT4-54) antibody has been tested by flow cytometric analysis of mouse resident peritoneal exudate cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative.
TIM-4 Monoclonal antibody specifically detects TIM-4 in Mouse samples. It is validated for Flow Cytometry.Specifications
TIM-4 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q6U7R4 | |
TIMD4 | |
Affinity chromatography | |
RUO | |
276891 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
54 (RMT4-54) | |
PerCP-eFluor 710 | |
TIMD4 | |
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.