Learn More
TIM-4 Monoclonal Antibody (54 (RMT4-54)), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£89.75 - £246.00
Specifications
Antigen | TIM-4 |
---|---|
Clone | 54 (RMT4-54) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15238859
|
Affymetrix eBioscience
12-5866-80 |
25 μg |
£89.75
25µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15248859
|
Affymetrix eBioscience
12-5866-82 |
100 μg |
£246.00
100µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody 54 recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC's and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells. Applications Reported: This 54 (RMT4-54) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 54 (RMT4-54) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Please consider the use of a viability dye like 7-AAD (cat 00-6993) when staining for Tim4. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.Specifications
TIM-4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 | |
TIMD4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TIMD4 |
54 (RMT4-54) | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
Q6U7R4 | |
276891 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.