missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
POA
Specifications
Clone | 344801 |
---|---|
Applications | Agonist Activity |
Classification | Monoclonal |
Target Species | Human |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Agonist Activity.Specifications
344801 | |
Monoclonal | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
IgG1 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects recombinant human TIM-3 in direct ELISAs and Western blots. |
Agonist Activity | |
Human | |
Q8TDQ0 | |
Mouse myeloma cell line NS0-derived recombinant human TIM-3 Met1-Arg200 Accession # Q8TDQ0 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only