missing translation for 'onlineSavingsMsg'
Learn More

TIM-3 Rat anti-Human, Clone: 344801, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB23651

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15364692

  • £275.00 / 100µg


Estimated Shipment
01-02-2018
Add to basket

Disclaimers:

For Research Use Only


Description

Description

TIM-3 Monoclonal antibody MAB23651 Specifically detects TIM-3 in Human samples. It is validated for Agonist Activity
Specifications

Specifications

TIM-3
344801
Supplied as a 0.2μm filtered solution in PBS.
Purified
Mouse myeloma cell line NS0-derived recombinant human TIM-3 ; AA Info: Met1-Arg200
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84868
Reconstitute at 0.5mg/mL in sterile PBS.
Agonist Activity
Unconjugated
Q8TDQ0
Rat
IgG1
100μg
-20 to -70°C as supplied; 1 month
Monoclonal
Detects recombinant human TIM-3 in direct ELISAs and Western blots.
Human
Videos