missing translation for 'onlineSavingsMsg'
Learn More

Tiglic acid, 97+%, Thermo Scientific Chemicals

Catalog Number p-7021533
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
80-59-1
C5H8O2
100.12
MFCD00066864
UIERETOOQGIECD-ONEGZZNKSA-N
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
125468
CHEBI:9592
(E)-2-methylbut-2-enoic acid
C\C=C(/C)C(O)=O
This item is not returnable. View return policy
80-59-1
C5H8O2
100.12
MFCD00066864
UIERETOOQGIECD-ONEGZZNKSA-N
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
125468
CHEBI:9592
(E)-2-methylbut-2-enoic acid
C\C=C(/C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 66°C
Boiling Point 198°C to 199°C
UN Number UN3261
Beilstein 1236500
Merck Index 149433
Formula Weight 100.12
Percent Purity ≥98%
Density 0.969
Quantity 25 g
Flash Point 101°C (213°F)
Chemical Name or Material Tiglic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'