missing translation for 'onlineSavingsMsg'
Learn More

Tianeptine sodium salt, Tocris Bioscience™

Selective facilitator of 5-HT uptake; antidepressant

Brand:  Tocris (Bio-Techne) 197210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460334

  • £147.15 / 10mg


Estimated Shipment
02-10-2018
Add to basket

Description

Description

Selective facilitator of 5-HT uptake in vitro and in vivo. Has no affinity for a wide range of receptors, including 5-HT and dopamine (IC50 > 10 μM) and has no effect on noradrenalin or dopamine uptake. Antidepressant, analgesic and neuroprotective following systemic administration in vivo.
Specifications

Chemical Identifiers

30123-17-2
458.933
7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt
C21H24ClN2NaO4S
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
23663953

Specifications

Tianeptine sodium salt
>99%
7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt
CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]
458.933
458.93
30123-17-2
C21H24ClN2NaO4S
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate
23663953
10mg
Videos