missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thymopoietin Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Thymopoietin/LAP2 |
---|---|
Clone | 60000000000 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Thymopoietin/LAP2 Monoclonal specifically detects Thymopoietin/LAP2 in Human samples. It is validated for Western Blot.Specifications
Thymopoietin/LAP2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TMPO, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma | |
TMPO | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
60000000000 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P42166 | |
7112 | |
E. coli-derived recombinant human Thymopoietin aa 2-187 Accession # P42166 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Thymopoietin in direct ELISAs and Western blots. |
For Research Use Only