missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
36.01 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
Description
Water soluble pH indicatorThymol Blue sodium salt acts as a pH indicator and utilized to indicate the changes in pH. It is involved in the ionization in multi-component drug formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼284°C (decomposition) |
Beilstein | 3861515 |
Solubility Information | Soluble in waterSoluble in 0;1N hydrochloric acid/ethanol. |
Formula Weight | 488.58 |
Grade | ACS |
Quantity | 10 g |
Physical Form | Crystalline |
Chemical Name or Material | Thymol Blue sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'