missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thymol Blue sodium salt, ACS

Water soluble pH indicator

Brand:  Alfa Aesar™ 042785.04

Code : NEW

Additional Details : CAS Number : 62625-21-2 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11315617

  • £9.80 / 2g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Thymol Blue sodium salt
ACS Grade
488.58
3861515
62625-21-2
C27H29NaO5S
MFCD00151093

Specifications

Thymol Blue sodium salt
ACS Grade
C27H29NaO5S
MFCD00151093
Soluble in water
2g
62625-21-2
Crystalline
488.58
3861515
≈284°C (dec.)
Videos
SDS