missing translation for 'onlineSavingsMsg'
Learn More

Thulium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7059198
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7440-30-4
Tm
168.93
MFCD00011281
FRNOGLGSGLTDKL-UHFFFAOYSA-N
tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
23961
CHEBI:33380
thulium
[Tm]
This item is not returnable. View return policy
7440-30-4
Tm
168.93
MFCD00011281
FRNOGLGSGLTDKL-UHFFFAOYSA-N
tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
23961
CHEBI:33380
thulium
[Tm]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Gray to Silver
UN Number UN3089
Quantity 1 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Thulium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'