missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thulium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific Chemicals
68.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Thulium(III) oxide when irradiated in a nuclear reactor, thulium produces an isotope that emits x-rays. A button of this isotope is used to make a lightweight, portable x-ray machine for medical use. It is also used in lasers with surgical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2425°C |
Color | Green |
Physical Form | Powder |
Quantity | 1 g |
Linear Formula | Tm2O3 |
Merck Index | 14,9394 |
Solubility Information | Insoluble in water. Slightly soluble in acids. |
Formula Weight | 385.87 |
Percent Purity | 99.997% |
Grade | Reacton™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'