Learn More
Thermo Scientific Alfa Aesar Thulium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)
Brand: Thermo Scientific Alfa Aesar 010376.FF
Thulium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)
Code : NEW
Additional Details : CAS Number : 7440-30-4 Weight : 0.01000kg Transport : UN number : 3178 Chem class : 4.1 Pack group : II