missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin-1 Mouse anti-Human, Mouse, Rat, Clone: A6.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9756-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15577636

  • £301.00 / 100µg

Estimated Shipment
04-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Specifications

Specifications

Thrombospondin-1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P35441, P07996
THBS, THBS-1, TSP, TSP-1, TSP1, thrombospondin-1, THBS1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
A6.1
Unconjugated
THBS1
Liquid
Thbs1
IgG1, kappa
100 μg
4° C
Primary
21825, 445442, 7057
Videos
SDS
Documents

For Research Use Only.