missing translation for 'onlineSavingsMsg'
Learn More

(±)-threo-3-Methylglutamic acid, Tocris Bioscience™

EAAT2 and EAAT4 blocker

Brand:  Tocris (Bio-Techne) 081150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458033

  • £294.37 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

A potent blocker of glutamate transport; selective for EAAT2 and EAAT4 (IC50 values are 90, 109, 1600 and 1080 μM for EAAT2, EAAT4, EAAT1 and EAAT3 respectively).
Specifications

Chemical Identifiers

63088-04-0
161.157
3-methylglutamic acid, 2s,3r-3-methylglutamic acid, 3r-3-methyl-l-glutamic acid, +/--threo-3-methylglutamic acid, threo-3-methyl-dl-glutamic acid, 3s-rel-3-methyl-d-glutamic acid, 2s,3r-2-azanyl-3-methyl-pentanedioic acid, inverted exclamation marka-threo-3-methylglutamic acid
(2S,3R)-2-amino-3-methylpentanedioic acid
C6H11NO4
FHJNAFIJPFGZRI-WUJLRWPWSA-N
11412540
CC(CC(=O)O)C(C(=O)O)N

Specifications

(±)-threo-3-Methylglutamic acid
C6H11NO4
FHJNAFIJPFGZRI-WUJLRWPWSA-N
(2S,3R)-2-amino-3-methylpentanedioic acid
11412540
>98%
63088-04-0
3-methylglutamic acid, 2s,3r-3-methylglutamic acid, 3r-3-methyl-l-glutamic acid, +/--threo-3-methylglutamic acid, threo-3-methyl-dl-glutamic acid, 3s-rel-3-methyl-d-glutamic acid, 2s,3r-2-azanyl-3-methyl-pentanedioic acid, inverted exclamation marka-threo-3-methylglutamic acid
CC(CC(=O)O)C(C(=O)O)N
161.157
161.16
50mg
Videos