Learn More
Invitrogen™ THRA Human Protein, Ligand Binding Domain, GST-Tagged
Thyroid Receptor alpha, Ligand Binding Domain (THRA LBD ) is a 57.90 kDa recombinant human protein (amino acids 148-410) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4762
Code : 50
Additional Details : Weight : 0.50000kg
Description
THRA LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
Store in ultra-cold freezer (-68 to -85°C). | |
Ligand Binding Domain | |
Human | |
Hormone Receptors |
50 μg | |
RUO - research use only | |
TR-α LBD | |
Recombinant |