Learn More
ThPOK Monoclonal Antibody (2POK), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£83.75 - £249.00
Specifications
Antigen | ThPOK |
---|---|
Clone | 2POK |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15228919
|
Affymetrix eBioscience
12-5928-82 |
100 μg |
£249.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15218919
|
Affymetrix eBioscience
12-5928-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This 2POK monoclonal antibody reacts with mouse ThPOK, which is also known as c-Krox and Zbtb7b. In hematopoietic cells, this 78 kDa transcription factor is expressed in thymocytes and T lymphocytes in the spleen and lymph nodes. In non-hematopoietic cells, ThPOK can be detected in the skin, heart, smooth muscle, and liver. Studies demonstrate that ThPOK promotes CD4+ lineage differentiation by inhibiting the expression of CD8 lineage-specific genes such as Runx3. ThPOK has also been shown to inhibit Cd4 silencing, another key facet required for differentiation to the CD4+ lineage. In addition to its role in thymocyte development, ThPOK is involved in the effector functions of invariant natural killer T (iNKT) cells and gamma delta T cells. Applications Reported: This 2POK antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 2POK antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.Specifications
ThPOK | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q64321 | |
Zbtb7b | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Zbtb7b |
2POK | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
22724 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.