missing translation for 'onlineSavingsMsg'
Learn More

Thorium Solution for ICP, 10g/L, HNO3 Matrix, Fisher Chemical™

Catalog Number p-7084008
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10±0.02g/L
Packaging Bottle
Quantity 100 mL
Type Mass Spectrometry Standard
Analyte or Component Names Th
Solvent or Matrix HNO3
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Thorium (Th) 10000mg/l in HNO3 5%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'