missing translation for 'onlineSavingsMsg'
Learn More

Thorin, For Spectrometric Determination, Honeywell™ Fluka™

Brand:  Honeywell Fluka 89120-1G

Code : NEW

Additional Details : CAS Number : 3688-92-4 Weight : 0.00100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 15637380

  • £93.17 / 1g


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Specifications

Chemical Identifiers

576.3g/mol
3688-92-4
MFCD00003888
Thorin
2957648
UN3465

Specifications

Thorin
Glass bottle
MFCD00003888
2957648
Dark red
1g
3688-92-4
576.3g/mol
UN3465
Soluble in water
6.1
Odorless
Videos