missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
22.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 175°C to 178°C |
Color | White |
Density | 1.405 g/mL |
Odor | Odorless |
Linear Formula | H2NCSNH2 |
Quantity | 50 g |
UN Number | UN3077 |
Beilstein | 605327 |
Merck Index | 14,9367 |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'