missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide, 98+%, Thermo Scientific Chemicals

Catalog Number 13408368
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
79-19-6
CH5N3S
91.13
MFCD00007620
BRWIZMBXBAOCCF-UHFFFAOYSA-N
thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
2723789
aminothiourea
C(=S)(N)NN
This item is not returnable. View return policy
79-19-6
CH5N3S
91.13
MFCD00007620
BRWIZMBXBAOCCF-UHFFFAOYSA-N
thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
2723789
aminothiourea
C(=S)(N)NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175.0°C to 180.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula H2NCSNHNH2
Quantity 2.5 kg
Beilstein 03,195
Fieser 01,1164
Merck Index 15,9514
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 91.13
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Thiosemicarbazide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Thiosemicarbazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'