missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiosemicarbazide, 98%, Thermo Scientific Chemicals
16.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
Solubility
Soluble in water, ethanol, alcohols and hot methanol.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
Solubility
Soluble in water, ethanol, alcohols and hot methanol.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | ∼180°C (decomposition) |
Odor | Odorless |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 506320 |
Merck Index | 14,9361 |
Solubility Information | Soluble in water,ethanol,alcohols and hot methanol. |
Formula Weight | 91.14 |
Percent Purity | 99% |
Chemical Name or Material | Thiosemicarbazide |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H300-Fatal if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'