missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, 99+%, Thermo Scientific Chemicals

Catalog Number p-7022928
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
108-98-5
C6H6S
110.174
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S
This item is not returnable. View return policy
108-98-5
C6H6S
110.174
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S

Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.

Notes
Incompatible with strong acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -15°C
Density 1.075
pH 5
Boiling Point 168°C to 169°C
Flash Point 55°C (131°F)
Assay Percent Range ≥99%
Odor Stench
Refractive Index 1.59
Quantity 50 g
UN Number UN2337
Beilstein 506523
Sensitivity Air Sensitive
Merck Index 14,9355
Solubility Information Miscible with most organic solvents and aqueous base. Immiscible with water.
Formula Weight 110.18
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'