missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99.5%, extra pure, benzene free, Thermo Scientific Chemicals

Catalog Number 10451022
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
110-02-1
C4H4S
84.14
MFCD00005413
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
8030
CHEBI:30856
thiophene
S1C=CC=C1
This item is not returnable. View return policy
110-02-1
C4H4S
84.14
MFCD00005413
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
8030
CHEBI:30856
thiophene
S1C=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -38.0°C
Density 1.0650g/mL
Boiling Point 84.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290
Quantity 100 mL
Beilstein 17, 29
Merck Index 15, 9506
Specific Gravity 1.065
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 84.14
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Thiophene, 99.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'