missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxylic hydrazide, 97%, Thermo Scientific Chemicals

Catalog Number p-7081336
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2361-27-5
C5H6N2OS
142.176
MFCD00005435
SOGBOGBTIKMGFS-UHFFFAOYSA-N
2-thiophenecarboxylic acid hydrazide, thiophene-2-carboxylic acid hydrazide, 2-thenoylhydrazine, 2-thiophenecarboxylic hydrazide, 2-thiophenecarbohydrazide, thiophene-2-carboxylic hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarbohydrazonic acid, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide
73098
thiophene-2-carbohydrazide
C1=CSC(=C1)C(=O)NN
This item is not returnable. View return policy
2361-27-5
C5H6N2OS
142.176
MFCD00005435
SOGBOGBTIKMGFS-UHFFFAOYSA-N
2-thiophenecarboxylic acid hydrazide, thiophene-2-carboxylic acid hydrazide, 2-thenoylhydrazine, 2-thiophenecarboxylic hydrazide, 2-thiophenecarbohydrazide, thiophene-2-carboxylic hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarbohydrazonic acid, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide
73098
thiophene-2-carbohydrazide
C1=CSC(=C1)C(=O)NN

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 137°C to 140°C
Quantity 5 g
Beilstein 114437
Formula Weight 142.18
Percent Purity ≥98%
Chemical Name or Material Thiophene-2-carboxylic hydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'