missing translation for 'onlineSavingsMsg'
Learn More

Thionicotinamide, 99%, Thermo Scientific Chemicals

Catalog Number p-200176
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4621-66-3
C6H6N2S
138.19
MFCD00006399
XQWBMZWDJAZPPX-UHFFFAOYSA-N
thionicotinamide, 3-pyridinecarbothioamide, nicotinothioamide, 3-thiocarbamoylpyridine, 3-thioamidopyridine, nicotinamide, thio, 3-pyridylthioformamide, 3-pyridinethiocarboxamide, thio-3-pyridinecarboxamide, unii-6a0hxx0753
737155
pyridine-3-carbothioamide
NC(=S)C1=CC=CN=C1
This item is not returnable. View return policy
4621-66-3
C6H6N2S
138.19
MFCD00006399
XQWBMZWDJAZPPX-UHFFFAOYSA-N
thionicotinamide, 3-pyridinecarbothioamide, nicotinothioamide, 3-thiocarbamoylpyridine, 3-thioamidopyridine, nicotinamide, thio, 3-pyridylthioformamide, 3-pyridinethiocarboxamide, thio-3-pyridinecarboxamide, unii-6a0hxx0753
737155
pyridine-3-carbothioamide
NC(=S)C1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 185°C to 190°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 0.2g/100 mL (20°C)
Formula Weight 138.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thionicotinamide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'