missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine, 97%, Thermo Scientific Chemicals

Catalog Number p-3593533
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
123-90-0
C4H9NS
103.19
MFCD00005974
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
thiomorpholine
C1CSCCN1
This item is not returnable. View return policy
123-90-0
C4H9NS
103.19
MFCD00005974
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
thiomorpholine
C1CSCCN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0880g/mL
Boiling Point 175°C to 178°C
Flash Point 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.539
Quantity 5 g
Beilstein 27, 9
Merck Index 15, 9452
Specific Gravity 1.088
Solubility Information Solubility in water: miscible. Other solubilities: miscible with many organic liquids
Formula Weight 103.19
Percent Purity 97%
Chemical Name or Material Thiomorpholine
Show More Show Less
compliance-icons
Product Identifier
  • Thiomorpholine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'