missing translation for 'onlineSavingsMsg'
Learn More

Thioisonicotinamide, 97%, ACROS Organics™

Brand:  Acros Organics 138770050

Code : 29

Additional Details : CAS Number : 2196-13-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10882631

  • £20.50 / 5g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Thioisonicotinamide
97%
C6H6N2S
MFCD00006437
2196-13-6
97%
138.19

Specifications

Thioisonicotinamide
198.0° to 202.0°C
2196-13-6
Glass bottle
97%
C6H6N2S
MFCD00006437
Cream to beige to yellow-brown
5g
Authentic
Crystalline Powder
97%
138.19
Videos
SDS