missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic acid, 98%, ACROS Organics™

£16.00 - £67.42

Chemical Identifiers

CAS 123-93-3
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004359
InChI Key UVZICZIVKIMRNE-UHFFFAOYSA-N
Synonym thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
PubChem CID 31277
ChEBI CHEBI:35012
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071650
SDS View Documents
Acros Organics
138740250
25g Plastic bottle
£16.00
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
10326450
SDS View Documents
Acros Organics
138741000
100g Plastic bottle
£20.67
100g
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
10071640
SDS View Documents
Acros Organics
138740010
1kg Plastic bottle
£67.42
1kg
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

123-93-3
150.148
UVZICZIVKIMRNE-UHFFFAOYSA-N
31277
2-(carboxymethylsulfanyl)acetic acid
C4H6O4S
MFCD00004359
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
CHEBI:35012
C(C(=O)O)SCC(=O)O

Specifications

Thiodiglycolic acid
123-93-3
C4H6O4S
MFCD00004359
15, 9484
UVZICZIVKIMRNE-UHFFFAOYSA-N
2-(carboxymethylsulfanyl)acetic acid
31277
150.15
98%
Plastic bottle
128.0°C to 131.0°C
25g
98%
98%
S(CH2CO2H)2
03, 253
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
C(C(=O)O)SCC(=O)O
150.148
CHEBI:35012
Crystalline Powder
Authentic
Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene
Gray to White
SDS