missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiocarbohydrazide, 98%, Thermo Scientific Chemicals
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 168.0°C to 172.0°C |
Color | Beige-Gray to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min |
Packaging | Glass bottle |
Linear Formula | H2NNHCSNHNH2 |
Quantity | 5 g |
Beilstein | 03, 197 |
Formula Weight | 106.15 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Thiocarbohydrazide
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Acute toxicity Category 3
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H300-Fatal if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'