missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
16.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 112°C to 114°C |
Quantity | 10 g |
Beilstein | 506006 |
Merck Index | 14,9319 |
Solubility Information | Soluble in ethanol,benzene and petroleum ether. Slightly soluble in water and ether. |
Formula Weight | 75.13 |
Percent Purity | 99% |
Grade | ACS |
Physical Form | Crystalline |
Chemical Name or Material | Thioacetamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'