missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, 98%, Thermo Scientific Chemicals

Catalog Number p-3346578
13.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-55-5
C2H5NS
75.13
MFCD00008070
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(N)=S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-55-5
C2H5NS
75.13
MFCD00008070
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(N)=S

Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

Solubility
Soluble in ethanol, benzene and petroleum ether. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 110°C to 114°C
Density 1.37
Assay Percent Range 98%
Quantity 25 g
Beilstein 506006
Merck Index 14,9319
Solubility Information Soluble in ethanol,benzene and petroleum ether; Slightly soluble in water and ether.
Formula Weight 75.13
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Thioacetamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'